Assay Details
| Gene Symbol: LOC613444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076273.1 | NP_001069741.1 | 2 - 3 | 529 | 105 | |
| GenBank mRNA | BC122670.1 | - | 2 - 3 | 529 | 105 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC613444
Genomic Map