Assay Details
| Gene Symbol: LOC613444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076273.1 | NP_001069741.1 | 7 - 8 | 1091 | 75 | |
| GenBank mRNA | BC122670.1 | - | 7 - 8 | 1091 | 75 | |
More Information
Set Membership: |
|

LOC613444
Genomic Map