Assay Details
| Gene Symbol: ARHGDIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076286.1 | NP_001069754.1 | 5 - 6 | 531 | 77 | |
| GenBank mRNA | BC122764.1 | - | 5 - 6 | 531 | 77 | |
More Information
Set Membership: |
|

ARHGDIG
Genomic Map