Assay Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076323.2 | NP_001069791.1 | 5 - 6 | 569 | 102 | |
| GenBank mRNA | BC120164.1 | - | 5 - 6 | 570 | 102 | |
More Information
Set Membership: |
|

DERL3
Genomic Map