Assay Details
| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076338.1 | NP_001069806.1 | 5 - 6 | 752 | 72 | |
| GenBank mRNA | BC120136.1 | - | 5 - 6 | 752 | 72 | |
More Information
Set Membership: |
|

MOB1A
Genomic Map