Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076376.1 | NP_001069844.1 | 2 - 3 | 204 | 66 | |
| GenBank mRNA | BC102300.1 | - | 2 - 3 | 204 | 66 | |
More Information
Set Membership: |
|

NME2
Genomic Map