Assay Details
| Gene Symbol: OCEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076386.2 | NP_001069854.2 | 3 - 4 | 387 | 62 | |
| GenBank mRNA | BC120466.1 | - | 3 - 4 | 387 | 62 | |
More Information
Set Membership: |
|

OCEL1
Genomic Map