Assay Details
| Gene Symbol: OPALIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076423.2 | NP_001069891.1 | 2 - 3 | 199 | 73 | |
| GenBank mRNA | BC120118.1 | - | 2 - 3 | 203 | 73 | |
More Information
Set Membership: |
|

OPALIN
Genomic Map