Assay Details
| Gene Symbol: KBTBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076463.1 | NP_001069931.1 | 1 - 2 | 32 | 62 | |
| GenBank mRNA | BC120403.1 | - | 1 - 2 | 32 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120403.1, NM_001076463.1 transcripts.
Set Membership: |
|

KBTBD4
Genomic Map