Assay Details
| Gene Symbol: HACD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076522.2 | NP_001069990.1 | 6 - 7 | 652 | 120 | |
| GenBank mRNA | BC120231.1 | - | 6 - 7 | 655 | 120 | |
More Information
Set Membership: |
|

HACD4
Genomic Map