Assay Details
| Gene Symbol: TMEM131L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076803.1 | NP_001070271.2 | 15 - 16 | 1573 | 68 | |
| GenBank mRNA | BC123545.1 | - | 15 - 16 | 1573 | 68 | |
More Information
Set Membership: |
|

TMEM131L
Genomic Map