Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076803.1 | NP_001070271.2 | 25 - 26 | 3415 | 144 | |
GenBank mRNA | BC123545.1 | - | 25 - 26 | 3415 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
