Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076803.1 | NP_001070271.2 | 5 - 6 | 462 | 95 | |
GenBank mRNA | BC123545.1 | - | 5 - 6 | 462 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123545.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
