Assay Details
| Gene Symbol: SPNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076808.2 | NP_001070276.1 | 6 - 7 | 1165 | 62 | |
| GenBank mRNA | BC123522.1 | - | 6 - 7 | 1167 | 62 | |
| BT030723.1 | - | 6 - 7 | 1154 | 62 | ||
More Information
Set Membership: |
|

SPNS1
Genomic Map