Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076816.1 | NP_001070284.1 | 2 - 3 | 102 | 58 | |
| GenBank mRNA | BC123758.1 | - | 2 - 3 | 102 | 58 | |
More Information
Set Membership: |
|

ZNF135
Genomic Map