Assay Details
| Gene Symbol: TM7SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076827.2 | NP_001070295.1 | 9 - 10 | 1304 | 59 | |
| GenBank mRNA | BC123528.1 | - | 9 - 10 | 1305 | 59 | |
More Information
Set Membership: |
|

TM7SF3
Genomic Map