Assay Details
| Gene Symbol: KCNN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076905.1 | NP_001070373.1 | 2 - 3 | 692 | 64 | |
| GenBank mRNA | BC123874.1 | - | 2 - 3 | 692 | 64 | |
More Information
Set Membership: |
|

KCNN4
Genomic Map