Assay Details
| Gene Symbol: RGS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076915.1 | NP_001070383.1 | 4 - 5 | 509 | 76 | |
| GenBank mRNA | BC123821.1 | - | 4 - 5 | 509 | 76 | |
More Information
Set Membership: |
|

RGS19
Genomic Map