Assay Details
| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076917.1 | NP_001070385.1 | 8 - 9 | 1463 | 96 | |
| GenBank mRNA | BC123881.1 | - | 8 - 9 | 1463 | 96 | |
More Information
Set Membership: |
|

KRT75
Genomic Map