Assay Details
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076943.1 | NP_001070411.1 | 5 - 6 | 709 | 135 | |
GenBank mRNA | BC119856.1 | - | 5 - 6 | 709 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
