Assay Details
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076963.1 | NP_001070431.1 | 2 - 3 | 109 | 146 | |
GenBank mRNA | BC120120.1 | - | 2 - 3 | 109 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120120.1, NM_001076963.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
