Assay Details
| Gene Symbol: ZSCAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077138.1 | NP_001070606.1 | 3 - 4 | 746 | 157 | |
| GenBank mRNA | BC123401.1 | - | 3 - 4 | 746 | 157 | |
More Information
Set Membership: |
|

ZSCAN12
Genomic Map