Assay Details
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077835.1 | NP_001071303.1 | 4 - 5 | 697 | 113 | |
GenBank mRNA | BC122603.1 | - | 4 - 5 | 697 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
