Assay Details
| Gene Symbol: SFTPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077838.2 | NP_001071306.2 | 3 - 4 | 350 | 142 | |
| GenBank mRNA | BC123546.1 | - | 3 - 4 | 351 | 142 | |
| EV634454.1 | - | 3 - 4 | 353 | 142 | ||
More Information
Set Membership: |
|

SFTPA1
Genomic Map