Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077871.1 | NP_001071339.1 | 3 - 4 | 445 | 53 | |
| GenBank mRNA | BC123695.1 | - | 3 - 4 | 445 | 53 | |
More Information
Set Membership: |
|

SLC25A29
Genomic Map