Assay Details
| Gene Symbol: EIF4EBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077893.2 | NP_001071361.1 | 2 - 3 | 341 | 65 | |
| GenBank mRNA | BC120290.1 | - | 2 - 3 | 342 | 65 | |
More Information
Set Membership: |
|

EIF4EBP1
Genomic Map