Assay Details
| Gene Symbol: CDKN1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077903.2 | NP_001071371.1 | 3 - 4 | 909 | 126 | |
| GenBank mRNA | BC123620.1 | - | 3 - 4 | 910 | 126 | |
More Information
Set Membership: |
|

CDKN1C
Genomic Map