Assay Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077913.2 | NP_001071381.1 | 1 - 2 | 87 | 85 | |
| GenBank mRNA | BC123592.1 | - | 1 - 2 | 88 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123592.1, NM_001077913.2 transcripts.
Set Membership: |
|

CHST12
Genomic Map