Assay Details
| Gene Symbol: AK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077933.2 | NP_001071401.1 | 3 - 4 | 564 | 90 | |
| GenBank mRNA | BC120077.1 | - | 3 - 4 | 566 | 90 | |
More Information
Set Membership: |
|

AK4
Genomic Map