Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078078.1 | NP_001071546.1 | 2 - 3 | 131 | 73 | |
GenBank mRNA | BC123607.1 | - | 2 - 3 | 131 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123607.1, NM_001078078.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
