Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078078.1 | NP_001071546.1 | 4 - 5 | 346 | 113 | |
GenBank mRNA | BC123607.1 | - | 4 - 5 | 346 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
