Assay Details
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078082.2 | NP_001071550.1 | 6 - 7 | 787 | 64 | |
GenBank mRNA | BC123604.1 | - | 6 - 7 | 788 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
