Assay Details
Gene Symbol: NR1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078100.2 | NP_001071568.1 | 7 - 8 | 2176 | 72 | |
GenBank mRNA | BC123488.1 | - | 7 - 8 | 1798 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
