Assay Details
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078131.1 | NP_001071599.1 | 2 - 3 | 281 | 86 | |
GenBank mRNA | BC123769.1 | - | 2 - 3 | 281 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
