Assay Details
Gene Symbol: DDIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078163.1 | NP_001071631.1 | 3 - 4 | 307 | 89 | |
GenBank mRNA | BC122721.1 | - | 3 - 4 | 307 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
