Assay Details
| Gene Symbol: MRPS30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079595.1 | NP_001073063.1 | 1 - 2 | 593 | 81 | |
| GenBank mRNA | BC122665.1 | - | 1 - 2 | 593 | 81 | |
More Information
Set Membership: |
|

MRPS30
Genomic Map