Assay Details
| Gene Symbol: MRPL39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080730.2 | NP_001074199.1 | 8 - 9 | 927 | 85 | |
| GenBank mRNA | BC122667.1 | - | 8 - 9 | 928 | 85 | |
More Information
Set Membership: |
|

MRPL39
Genomic Map