Assay Details
| Gene Symbol: AMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173990.2 | NP_776415.1 | 8 - 9 | 1171 | 73 | |
| GenBank mRNA | BC123795.1 | - | 8 - 9 | 1171 | 73 | |
| M95605.1 | - | 8 - 9 | 1170 | 73 | ||
More Information
Set Membership: |
|

AMD1
Genomic Map