Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174065.2 | NP_776490.2 | 8 - 9 | 1244 | 58 | |
| GenBank mRNA | BC119892.1 | - | 8 - 9 | 1244 | 58 | |
More Information
Set Membership: |
|

GFAP
Genomic Map