Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174071.2 | NP_776496.1 | 8 - 9 | 979 | 59 | |
| GenBank mRNA | AF268376.1 | - | 7 - 8 | 1022 | 59 | |
| BC123491.1 | - | 8 - 9 | 979 | 59 | ||
More Information
Set Membership: |
|

GLRB
Genomic Map