Assay Details
| Gene Symbol: M6PR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175779.3 | NP_786973.1 | 6 - 7 | 874 | 65 | |
| GenBank mRNA | BC120081.1 | - | 6 - 7 | 874 | 65 | |
| BT029887.1 | - | 6 - 7 | 839 | 65 | ||
| M17025.1 | - | 6 - 7 | 754 | 65 | ||
More Information
Set Membership: |
|

M6PR
Genomic Map