Assay Details
Gene Symbol: AURKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183084.2 | NP_898907.2 | 4 - 5 | 519 | 80 | |
GenBank mRNA | AY336975.1 | - | 4 - 5 | 461 | 80 | |
BC123650.1 | - | 4 - 5 | 519 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
