Assay Details
| Gene Symbol: SAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203323.3 | NP_976068.1 | 5 - 6 | 615 | 74 | |
| GenBank mRNA | BC122800.1 | - | 5 - 6 | 616 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAT2| Gene Symbol: SAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203323.3 | NP_976068.1 | 5 - 6 | 615 | 74 | |
| GenBank mRNA | BC122800.1 | - | 5 - 6 | 616 | 74 | |
Set Membership: |
|