Assay Details
| Gene Symbol: PAOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013602.2 | NP_001013620.2 | 6 - 7 | 1397 | 60 | |
| GenBank mRNA | AF226658.1 | - | 5 - 6 | 1214 | 60 | |
| BT021621.1 | - | 7 - 8 | 1554 | 60 | ||
| DQ058607.1 | - | 6 - 7 | 1397 | 60 | ||
More Information
Set Membership: |
|

PAOX
Genomic Map