Assay Details
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015513.2 | NP_001015513.2 | 2 - 3 | 142 | 134 | |
GenBank mRNA | BC126571.1 | - | 2 - 3 | 143 | 134 | |
BT021657.1 | - | 2 - 3 | 130 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126571.1, NM_001015513.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
