Assay Details
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015513.2 | NP_001015513.2 | 3 - 4 | 188 | 114 | |
GenBank mRNA | BC126571.1 | - | 3 - 4 | 189 | 114 | |
BT021657.1 | - | 3 - 4 | 176 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
