Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017948.3 | NP_001017948.1 | 5 - 6 | 927 | 61 | |
| GenBank mRNA | BC133562.1 | - | 5 - 6 | 928 | 61 | |
| BT021886.1 | - | 5 - 6 | 827 | 61 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map