Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037141.3 | NP_001032218.2 | 2 - 3 | 203 | 54 | |
NM_001110194.1 | NP_001103664.1 | 5 - 6 | 604 | 54 | ||
GenBank mRNA | DQ219802.1 | - | 2 - 3 | 205 | 54 | |
DT721910.1 | - | 1 - 2 | 63 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
