Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045881.2 | NP_001039346.2 | 16 - 17 | 2724 | 83 | |
| GenBank mRNA | AY693782.1 | - | 7 - 8 | 689 | 83 | |
| BC105482.1 | - | 16 - 17 | 2726 | 83 | ||
More Information
Set Membership: |
|

CENPC
Genomic Map