Assay Details
Gene Symbol: LNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076234.2 | NP_001069702.2 | 6 - 7 | 1590 | 54 | |
GenBank mRNA | BC123654.1 | - | 5 - 6 | 1287 | 54 | |
BC134476.1 | - | 6 - 7 | 1590 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
